missing translation for 'onlineSavingsMsg'
Learn More

Ethylenebis(oxyethylenenitrilo)tetraacetic acid, 98%

Product Code. 10341591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10341591 25 g Plastic bottle 25g
10351591 100 g Plastic bottle 100g
10072823 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10341591 Supplier Thermo Scientific Acros Supplier No. 409910250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-42-5 | C14H24N2O10 | 380.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.34
MDL Number MFCD00004291
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
PubChem CID 6207
ChEBI CHEBI:30740
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Melting Point 240.0°C to 245.0°C
Color White
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula [-CH2OCH2CH2N(CH2CO2H)2]2
Quantity 25 g
Beilstein 04, IV, 2433
Merck Index 15, 3576
Solubility Information Solubility in water: slightly soluble
Formula Weight 380.34
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethylenebis(oxyethylenenitrilo)tetraacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.