missing translation for 'onlineSavingsMsg'
Learn More

Ethylenebistetrabromophthalimide, TRC

Product Code. 30133890
Change view
Click to view available options
Quantity:
500 mg
2.5 g
25 g
Unit Size:
2.5g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133890 25 g 25g
30007220 2.5 g 2.5g
30009535 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133890 Supplier Toronto Research Chemicals Supplier No. TRCE91730025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32588-76-4
Molecular Formula C18 H4 Br8 N2 O4
Molecular Weight (g/mol) 951.47
Synonym Phthalimide, N,N'-ethylenebis[tetrabromo- (8CI), 2,2'-(1,2-Ethanediyl)bis[4,5,6,7-tetrabromo-1H-isoindole-1,3(2H)-dione], 1,2-Bis(tetrabromophthalimido)ethane, BT 93, BT 93W, BT 93WFG, Chemguard 952, Citex BT 93, EcoFlame B 951, N,N'-Ethylenebis(tetrabromophthalimide), Saytex BT 93, Saytex BT 93W, Saytex BT 93W1
IUPAC Name 4,5,6,7-tetrabromo-2-[2-(4,5,6,7-tetrabromo-1,3-dioxoisoindol-2-yl)ethyl]isoindole-1,3-dione
SMILES Brc1c(Br)c(Br)c2C(=O)N(CCN3C(=O)c4c(Br)c(Br)c(Br)c(Br)c4C3=O)C(=O)c2c1Br

Specifications

Quantity 25 g
InChI Formula InChI=1S/C18H4Br8N2O4/c19-7-3-4(8(20)12(24)11(7)23)16(30)27(15(3)29)1-2-28-17(31)5-6(18(28)32)10(22)14(26)13(25)9(5)21/h1-2H2
Formula Weight 943.3638
Chemical Name or Material Ethylenebistetrabromophthalimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.