missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine, 99%

Product Code. p-3737935 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
5mL
This item is not returnable. View return policy

Product Code. 10244960

Brand: Thermo Scientific Acros 118420050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 107-15-3 | C2H8N2 | 60.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-15-3
Molecular Formula C2H8N2
Molecular Weight (g/mol) 60.10
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

Color Colorless to Yellow
Density 0.8980g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H2NCH2CH2NH2
Refractive Index 1.4555 to 1.4575 (20°C, 589nm)
Quantity 5 mL
Beilstein 04, 230
Fieser 01,372; 04,231; 13,157
Merck Index 15, 3849
Formula Weight 60.1
Percent Purity ≥98.5%
Grade Extra Pure
Physical Form Liquid, Fuming In Air
Chemical Name or Material Ethylenediamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.