missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Certified AR for Analysis, Fisher Chemical™

Product Code. 10786281 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10786281 100 g 100g
10479740 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10786281 Supplier Fisher Chemical Supplier No. D/0452/48
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number 3541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point 220°C
Color White
pH 2.5
Physical Form Powder/Solid
Quantity 100 g
Formula Weight 292.23g/mol
Packaging Plastic powder jar
Vapor Pressure 0.013 hPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.