missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Di K Salt Dihydrate (White Crystalline Powd.), Fisher BioReagents

Product Code. 10336363
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336363 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10336363 Supplier Fisher BioReagents Supplier No. BP119500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
The dipotassium salt of EDTA is used in place of the disodium salt where sodium would interfere with the reaction.
TRUSTED_SUSTAINABILITY

Specifications

CAS 25102-12-9
Melting Point 272°C
Color White
pH 4.0 to 5.0
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Molecular Formula C10H18K2N2O10
Linear Formula (HOOCCH2)2NCH2CH2N(CH2COOK)2·2H2O
Quantity 500 g
Synonym ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx
InChI Key JFROQFOCFOKDKU-UHFFFAOYSA-L
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+]
IUPAC Name dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Molecular Weight (g/mol) 404.455
PubChem CID 53486451
Formula Weight 404.47
Percent Purity ≥99.0%
Physical Form Powder or Solid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, Dipotassium Salt Dihydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, dipotassium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.