missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate, 97%

Product Code. 15711759 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15711759 500 g 500g
15791749 25 g 25g
15701759 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15711759 Supplier Thermo Scientific Acros Supplier No. 457935000
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 194491-32-2 | C10H12MgN2Na2O8 | 358.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194491-32-2

Specifications

Assay Percent Range 96.0 % min. (on dry substance)
Molecular Formula C10H12MgN2Na2O8·xH2O
Quantity 500 g
Solubility Information Solubility aq. soln.: soluble.
Molecular Weight (g/mol) 358.5
Formula Weight 358.5
Chemical Name or Material Ethylenediaminetetraacetic acid disodium magnesium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.