missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt concentrate with zinc complex added (solution B), For 1L standard solution, for complexometry, Solstice

Product Code. 15635200
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15635200 1 Ea. Each
15645200 6 x 1 Ea. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15635200 Supplier Honeywell-Fluka Supplier No. 380561EA
  • ONLINE EXCLUSIVE PRICE
    67.05 €
    63.07 € / Pack of 1
    Save 3.98 €
    5% Off
 Restricted Item
Estimated Shipment: 07-05-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution, 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-33-3 , 14025-21-9 , 64-02-8
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
PubChem CID 57339238
ChEBI CHEBI:64734
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 15.84, 1.19, 6.41
Physical Form Liquid
Quantity 1 Ea.
UN Number NONH for all modes of transport
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt concentrate with zinc complex added (solution B)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.