missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, for Electrophoresis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
100g
500g
5kg
Specifications
Specifications
| Melting Point | 252°C |
| Color | White |
| pH | 4 to 6 |
| Physical Form | Solid |
| Quantity | 500 g |
| Formula Weight | 372.23g/mol |
| Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction