missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt dihydrate, Reag. Ph. Eur., 99-101%, ≤0.1% NTA (HPLC), for complexometry, Solstice

Product Code. 15604270 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
6 x 50 g
1 kg
6 x 100 g
25 kg
6 x 1 kg
Unit Size:
100g
1kg
25kg
500g
50g
Pack of 6
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15604270 1 kg 1kg
15634270 50 g 50g
15694260 100 g 100g
15664270 6 x 50 g Pack of 6
15624270 500 g 500g
15644270 6 x 100 g Pack of 6
15654270 6 x 1 kg Pack of 6
15614270 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15604270 Supplier Honeywell-Fluka Supplier No. 345491KG
  • ONLINE EXCLUSIVE PRICE
    300.00 €
    281.99 € / 1kg
    Save 18.01 €
    6% Off
 Restricted Item
Estimated Shipment: 01-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reag. Ph. Eur., 99-101%, ≤0.1% NTA (HPLC), for complexometry

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
UN Number NONH for all modes of transport
Beilstein 3900609
Flash Point >100°C (>212°F)
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
Formula Weight 372.24g/mol
Percent Purity 99 to 101%
Grade Not Available
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt dihydrate
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid disodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.