missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Solution, 0.01M, Solstice

Product Code. 15684930
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15684930 500 mL 500mL
15674930 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15684930 Supplier Honeywell-Fluka Supplier No. 35322500ML
  • ONLINE EXCLUSIVE PRICE
    85.65 €
    77.45 € / 500mL
    Save 8.20 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 0.01 M EDTA-Na2, for complexometry

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
PubChem CID 57339238
ChEBI CHEBI:64734
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Physical Form Liquid
Quantity 500 mL
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
UN Number NONH for all modes of transport
Beilstein 3822669
Formula Weight 336.21g/mol
Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.