missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ EDTA disodium salt solution with zinc complex added (solution A), Volumetric, for water hardness determination, Honeywell Fluka™

Product Code. 15674260 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 X 1 L
Unit Size:
1 L
Pack of 6
Product Code. Quantity unitSize
15674260 1 L 1 L
15684260 6 X 1 L Pack of 6
2 options
This item is not returnable. View return policy

Product Code. 15674260

Brand: Honeywell Fluka™ 345471L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Volumetric, for water hardness determination (1 ml = 5.6 German degree of hardness in 100 ml of water)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 L
Type Ethylenediaminetetraacetic acid disodium salt solution with zinc complex added (solution A)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.