missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt, TRC

Product Code. 30010604
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010604 25 g 25g
30137908 250 g 250g
30136458 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010604 Supplier Toronto Research Chemicals Supplier No. TRCE91833525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10 H14 N2 O8 . 2Na
Molecular Weight (g/mol) 336.206
Synonym Glycine, N,N'-1,2-ethanediylbis[N-(carboxymethyl)-, sodium salt (1:2), Acetic acid, (ethylenedinitrilo)tetra-, disodium salt (8CI), Glycine, N,N'-1,2-ethanediylbis[N-(carboxymethyl)-, disodium salt (9CI), Chelaplex, Chelaplex III, Chelaton III, Chelest 200, Chelest 2BS, Chelest B, Clewat N, Clewat N 2, Complexon III, Disodium (ethylenedinitrilo)tetraacetate, Disodium EDTA, Disodium diacid ethylenediaminetetraacetate, Disodium dihydrogen ethylenediaminetetraacetate, Disodium edathamil, Disodium edetate, Disodium ethylenediamine-N,N,N',N'-tetraacetate, Disodium ethylenediaminetetraacetate, Disodium ethylenediaminetetraacetic acid salt, Disodium sequestrene, Disodium tetracemate, Disodium versenate, Disodium versene, Dissolvine Na 2T, Dissolvine Na 2x, Dissolvine Na2, Dissolvine Na2S, Dotite 2NA, EDTA disodium salt, Edathamil disodium, Edeta BD, Edetate Disodium, Endrate disodium, Ethylenediaminetetraacetic acid disodium salt, Ethylenedinitrilotetraacetate disodium, F 1, F 1 (complexon), Idranal III, Khelaton 3, Komplexon III, Mavacid ED 4, Metaquest B, Nervanaid Ba2, Perma Kleer Di Crystals, Salina, Selekton B2, Sequestrene Na2, Sodium versenate, TW 204, Tetracemate disodium, Titriplex III, Trilon BD, Triplex III, VRP 1034, Versene NA, Versene Na2, Versene disodium salt, Versonol 120, Zonon D
IUPAC Name disodium;2-[2-[carboxylatomethyl(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetate
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(=O)O)CC(=O)[O-])CC(=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H16N2O8.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;/q;2*+1/p-2
Formula Weight 336.0546
Percent Purity >95%
Chemical Name or Material Disodium Edetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.