missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure

Product Code. 15748687 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Nalgene bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15748687 500 mL Nalgene bottle 500mL
15825388 100 mL Nalgene bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15748687 Supplier Thermo Scientific Supplier No. J15694.AP
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chelating agent used in many enzyme buffers and at higher concentrations, as enzyme inactivator | CAS: 6381-92-6 | C10H18N2Na2O10 | 372.238 g/mol

Chelating agent used in many enzyme buffers and, at higher concentrations, as an enzyme inactivator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.238
MDL Number MFCD00150037
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]

Specifications

Packaging Nalgene bottle
Quantity 500 mL
Beilstein 3900609
Grade Molecular Biology, Ultrapure
Concentration or Composition (by Analyte or Components) 0.5 M
Physical Form Solution
Chemical Name or Material Ethylenediaminetetraacetic acid
compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, 0.5M soln.
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Disodium EDTA, Sodium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.