missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Electrophoresis Grade, 5kg, MP Biomedicals

Product Code. 11471562 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471562 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471562 Supplier MP Biomedicals Supplier No. 04800684
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

60-00-4 (EDTA) Purity: ≥99.5% Heavy metals (as Pb): <<10 ppm Iron (Fe): <<10 ppm Sulfated ash: <<0.1%

Specifications

CAS 60-00-4
Molecular Formula C10H16N2O8
MDL Number MFCD00003541
Quantity 5 kg
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol) 292.24
PubChem CID 6049
ChEBI CHEBI:42191
Chemical Name or Material Ethylenediaminetetraacetic Acid
Show More Show Less
compliance-icons
Product Identifier
  • Ethylenediamine Tetraacetic Acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ethylenediamine Tetraacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.