Learn More
Ethylenediaminetetraacetic acid tripotassium salt dihydrate, 99%
Description
Ethylenediaminetetraacetic acid tripotassium salt dihydrate is involved to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals. It is also used to study chelators, biochemicals and reagents. Further, it is utilized to check the safety of ethylenediaminetetraacetic acid and hydroxyethyenediaminetriacetic acid ingredients in cosmetic formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethylenediaminetetraacetic acid tripotassium salt dihydrate is involved to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals. It is also used to study chelators, biochemicals and reagents. Further, it is utilized to check the safety of ethylenediaminetetraacetic acid and hydroxyethyenediaminetriacetic acid ingredients in cosmetic formulations.
Solubility
Soluble in water and potassium hydroxide.
Notes
Incompatible with strong oxidizing agents, metals, zinc, aluminum, carbon steel, copper, copper alloys, galvanized metals and nickel.
Specifications
Specifications
| Melting Point | ∽182°C (decomposition) |
| Quantity | 250 g |
| Beilstein | 6762497 |
| Solubility Information | Soluble in water and potassium hydroxide. |
| Formula Weight | 442.55 (406.46 Anhydrous) |
| Percent Purity | 99% |
| Chemical Name or Material | Ethylenediaminetetraacetic acid tripotassium salt dihydrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.