missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate, MP Biomedicals™

Product Code. 11416882 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416882 1 kg 1kg
11406882 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11416882 Supplier MP Biomedicals Supplier No. 0215108891
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Water soluble. Clear colorless to light yellow solution at 32gm plus 100mL of 1M sodium hydroxide

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85715-60-2
Molecular Formula C10H18N2Na3O9
Molecular Weight (g/mol) 379.228
MDL Number MFCD00149675
InChI Key DFCJTQJARZUURB-UHFFFAOYSA-N
Synonym edta, trisodium salt trihydrate
PubChem CID 131865091
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;hydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.[Na].[Na].[Na]

Specifications

Color White
Quantity 1 kg
Formula Weight 358.19
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.