missing translation for 'onlineSavingsMsg'
Learn More

Ethylicin, TRC

Product Code. 30385632
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385632 50 mg -
30373800 100 mg -
30374801 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385632 Supplier Toronto Research Chemicals Supplier No. TRCE93778050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 682-91-7
Molecular Formula C4 H10 O2 S2
Molecular Weight (g/mol) 154.25
Synonym Ethanesulfonothioic acid, S-ethyl ester, Ethanesulfonic acid, thio-, S-ethyl ester (6CI,7CI,8CI), Ethanethiol, ethanesulfonate (6CI,8CI), Ethanethiosulfonic acid ethyl ester, Ethyl ethanethiosulfonate, NSC 22422, Pseudoallicin 150, S-Ethyl ethanethiosulfonate, Ethylicin
IUPAC Name 1-ethylsulfonylsulfanylethane
SMILES CCSS(=O)(=O)CC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H10O2S2/c1-3-7-8(5,6)4-2/h3-4H2,1-2H3
Formula Weight 154.0122
Chemical Name or Material Ethylicin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.