missing translation for 'onlineSavingsMsg'
Learn More

Ethylidenecyclopentane, 90+%, Thermo Scientific Chemicals

Product Code. 11479656
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479656 5 g 5g
11489656 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479656 Supplier Thermo Scientific Alfa Aesar Supplier No. B20209.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

It is used to make other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to make other organic chemicals.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2146-37-4
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00019319
InChI Key VONKRKBGTZDZNV-UHFFFAOYSA-N
Synonym cyclopentane, ethylidene, ethyl idenecyclopentane, ethylidene-cyclopentane, cyclopentane,ethylidene, acmc-1cs2b, 1e-ethylidenecyclopentane, ethylidenecyclopentane, 90+%
PubChem CID 75068
IUPAC Name ethylidenecyclopentane
SMILES CC=C1CCCC1

Specifications

Melting Point -130°C
Density 0.8
Boiling Point 112°C to 114°C
Refractive Index 1.45
Quantity 5 g
UN Number UN3295
Solubility Information Soluble in water.
Formula Weight 96.17
Percent Purity ≥90%
Chemical Name or Material Ethylidenecyclopentane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.