missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%

Product Code. 10020920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10020920 5 g Glass bottle 5g
10030920 25 g Glass bottle 25g
10336460 100 g Glass bottle 100g
10033313 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10020920 Supplier Thermo Scientific Chemicals Supplier No. 118680050
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

Ethylmercurithiosalicylic acid, CAS # 54-64-8, is an oxidizing reagent that can inhibit various sulfhydryl-dependent enzymes and proteins. It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt. | CAS: 54-64-8 | C9H9HgNaO2S | 404.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ethylmercurithiosalicylic acid is an oxidizing reagent that inhibits sulfhydryl-containing active sites of several enzymes
  • It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt.
  • It can bind to sulfhydryl compounds, such as glutathione, cysteine, and sulfhydryl groups of proteins
  • Additionally, it can activate inositol triphosphate calcium channels on membranes of the endoplasmic reticulum, resulting in the regulation of cellular functions

Applications

  • Ethylmercurithiosalicylic acid has been widely investigated as a preservative in medicinal laboratory research, including vaccine studies
  • It has been shown to exhibit antifungal and antiseptic properties
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym thimerosal, thiomersal, thiomersalate, mercurothiolate, sodium merthiolate, sodium ethylmercurithiosalicylate, merthiolate, thimerosalate, thimerosalum, thimersalate
PubChem CID 16684434
ChEBI CHEBI:9546
IUPAC Name sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Melting Point 232.0°C to 233.0°C
Color Cream to White
pH 6 to 8 (1% soln.)
Physical Form Powder
Quantity 5 g
Assay Percent Range 97.0-101.0%
Linear Formula 2-(C2H5HgS)C6H4CO2Na
Flash Point 250°C
Merck Index 15, 9470
Solubility Information Solubility in water: 1g/1mL (20°C). Other solubilities: 1g/8mL in alcohol,1g/10mL in methanol,almost insoluble in ether and benzene
Formula Weight 404.8
Percent Purity 97.0 to 101%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (vacuum/P2O5)
Packaging Glass bottle
Chemical Name or Material Ethylmercurithiosalicylic acid, sodium salt
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylmercurithiosalicylic acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sodium o-(ethylmercurithio)benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.