missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%

Product Code. 10336460 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10336460

Brand: Thermo Scientific Chemicals 118681000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ethylmercurithiosalicylic acid, CAS # 54-64-8, is an oxidizing reagent that can inhibit various sulfhydryl-dependent enzymes and proteins. It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt. | CAS: 54-64-8 | C9H9HgNaO2S | 404.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ethylmercurithiosalicylic acid is an oxidizing reagent that inhibits sulfhydryl-containing active sites of several enzymes
  • It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt.
  • It can bind to sulfhydryl compounds, such as glutathione, cysteine, and sulfhydryl groups of proteins
  • Additionally, it can activate inositol triphosphate calcium channels on membranes of the endoplasmic reticulum, resulting in the regulation of cellular functions

Applications

  • Ethylmercurithiosalicylic acid has been widely investigated as a preservative in medicinal laboratory research, including vaccine studies
  • It has been shown to exhibit antifungal and antiseptic properties
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-64-8
Molecular Formula C9H9HgNaO2S
Molecular Weight (g/mol) 404.81
MDL Number MFCD00013062
InChI Key RTKIYNMVFMVABJ-UHFFFAOYSA-L
Synonym thimerosal, thiomersal, thiomersalate, mercurothiolate, sodium merthiolate, sodium ethylmercurithiosalicylate, merthiolate, thimerosalate, thimerosalum, thimersalate
PubChem CID 16684434
ChEBI CHEBI:9546
IUPAC Name sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
SMILES [Na+].CC[Hg]SC1=CC=CC=C1C([O-])=O

Specifications

Melting Point 232.0°C to 233.0°C
Color Cream to White
pH 6 to 8 (1% soln.)
Physical Form Powder
Quantity 100 g
Assay Percent Range 97.0-101.0%
Linear Formula 2-(C2H5HgS)C6H4CO2Na
Flash Point 250°C
Merck Index 15, 9470
Solubility Information Solubility in water: 1g/1mL (20°C). Other solubilities: 1g/8mL in alcohol,1g/10mL in methanol,almost insoluble in ether and benzene
Formula Weight 404.8
Percent Purity 97.0 to 101%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (vacuum/P2O5)
Packaging Glass bottle
Chemical Name or Material Ethylmercurithiosalicylic acid, sodium salt
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylmercurithiosalicylic acid, sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium o-(ethylmercurithio)benzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.