missing translation for 'onlineSavingsMsg'
Learn More

(Ethylthio)acetone, 95%

Product Code. 11386806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11386806 25 g 25g
11376806 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386806 Supplier Thermo Scientific Alfa Aesar Supplier No. L04142.14
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 20996-62-7 | C5H10OS | 118.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20996-62-7
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00026976
InChI Key ZKDXKBIOEVBFGV-UHFFFAOYSA-N
PubChem CID 88750
IUPAC Name 1-ethylsulfanylpropan-2-one
SMILES CCSCC(=O)C

Specifications

Boiling Point 170°C to 172°C
Flash Point 52°C (125°F)
Refractive Index 1.472
Quantity 25 g
UN Number UN1993
Beilstein 1740830
Synonym ethylthio acetone, 2-propanone, 1-ethylthio, 1-ethylthio acetone, ethyl 2-oxopropyl sulfide, 1-ethylthio-2-propanone, 2-propanone,1-ethylthio, 1-ethylsulfanyl propan-2-one, thioethoxy acetone, acmc-1cazk, ch3coch2sch2ch3
Formula Weight 118.2
Percent Purity 95%
Chemical Name or Material (Ethylthio)acetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.