missing translation for 'onlineSavingsMsg'
Learn More

(Ethylthio)trimethylsilane, 90%, Thermo Scientific Chemicals

Product Code. 10347711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10347711 1 g Glass bottle 1g
10153251 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347711 Supplier Thermo Scientific Acros Supplier No. 345000010
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5573-62-6
Molecular Formula C5H14SSi
Molecular Weight (g/mol) 134.31
MDL Number MFCD00042890
InChI Key HXAFQWICVBBXMZ-UHFFFAOYSA-N
Synonym ethylthio trimethylsilane, ethylthiotrimethylsilane, ethylthio trimethylsilane, technical grade, acmc-20aplk, trimethyl ethylthio silane, ethyl thio trimethylsilane, ethyl trimethylsilyl sulfide, trimethylsilyl ethyl sulfide, ethylsulfanyl trimethylsilane, ethylsulfanyl trimethyl silane
PubChem CID 2733426
IUPAC Name ethylsulfanyl(trimethyl)silane
SMILES CCS[Si](C)(C)C

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless
Density 0.8300g/mL
Boiling Point 69°C to 70°C (100.0 mmHg)
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C2H5SSi(CH3)3
Refractive Index 1.4470 to 1.451
Quantity 1 g
Specific Gravity 0.83
Formula Weight 134.3
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material (Ethylthio)trimethylsilane, 90%
Show More Show Less
compliance-icons
Product Identifier
  • (Ethylthio)trimethylsilane, tech.
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.