missing translation for 'onlineSavingsMsg'
Learn More

Ethyltrichlorosilane, 99%, AcroSeal™, ACROS Organics™

Product Code. 10506651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10506651 100 mL Glass Bottle 100mL
11935841 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10506651 Supplier Thermo Scientific Acros Supplier No. 313511000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 115-21-9
Molecular Formula C2H5Cl3Si
Molecular Weight (g/mol) 163.497
InChI Key ZOYFEXPFPVDYIS-UHFFFAOYSA-N
Synonym ethyltrichlorosilane, trichloro ethyl silane, trichloroethylsilicane, silane, trichloroethyl, ethyl silicon trichloride, trichloroethylsilicon, silicane, trichloroethyl, ethyl trichlorosilane, silane, ethyltrichloro, c2h5sicl3
PubChem CID 8260
IUPAC Name trichloro(ethyl)silane
SMILES CC[Si](Cl)(Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.2380g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H5C2SiCl3
Refractive Index 1.4240 to 1.4260 (20°C, 589nm)
Quantity 100 mL
Formula Weight 163.51
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Ethyltrichlorosilane, 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.