missing translation for 'onlineSavingsMsg'
Learn More

Ethynyltri-n-butyltin, 96%

Product Code. 11339006
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339006 1 g 1g
11349006 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339006 Supplier Thermo Scientific Alfa Aesar Supplier No. L20178.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 994-89-8 | C14H28Sn | 315.088 g/mol

Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

Solubility
Not miscible or difficult to mix with water.

Notes
Mositure sensitive. Store away from oxidizing agents and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-89-8
Molecular Formula C14H28Sn
Molecular Weight (g/mol) 315.088
MDL Number MFCD00009420
InChI Key YEMJHNYABQHWHL-UHFFFAOYSA-N
Synonym tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
PubChem CID 621176
IUPAC Name tributyl(ethynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#C

Specifications

Density 1.08
Boiling Point 70°C (0.2 mmHg)
Flash Point 73°C (163°F)
Linear Formula HC≡C-Sn(CH2CH2CH2CH3)3
Refractive Index 1.477
Quantity 1 g
UN Number UN2788
Beilstein 3537659
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 315.09
Percent Purity 96%
Chemical Name or Material Ethynyltri-n-butyltin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.