missing translation for 'onlineSavingsMsg'
Learn More

Ethynyltri-n-butyltin, 96%

Product Code. 11349006
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349006 5 g 5g
11339006 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349006 Supplier Thermo Scientific Alfa Aesar Supplier No. L20178.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 994-89-8 | C14H28Sn | 315.088 g/mol

Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

Solubility
Not miscible or difficult to mix with water.

Notes
Mositure sensitive. Store away from oxidizing agents and water/moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-89-8
Molecular Formula C14H28Sn
Molecular Weight (g/mol) 315.088
MDL Number MFCD00009420
InChI Key YEMJHNYABQHWHL-UHFFFAOYSA-N
Synonym tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
PubChem CID 621176
IUPAC Name tributyl(ethynyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C#C

Specifications

Density 1.08
Boiling Point 70°C (0.2 mmHg)
Flash Point 73°C (163°F)
Linear Formula HC≡C-Sn(CH2CH2CH2CH3)3
Refractive Index 1.477
Quantity 5 g
UN Number UN2788
Beilstein 3537659
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 315.09
Percent Purity 96%
Chemical Name or Material Ethynyltri-n-butyltin
Show More Show Less

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.