missing translation for 'onlineSavingsMsg'
Learn More

Etomidate, MedChemExpress

Product Code. 16478338
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478338 10 mg 10mg
16468086 100 mg 100mg
16454227 50 mg 50mg
16421376 200 mg 200mg
16402016 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16478338 Supplier Medchem Express Supplier No. HYB0100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Etomidate (R 16659) is a potent GABAA receptor agonist. Etomidate is a neurological drug and effective parenteral medication and has the potential for management of endogenous hypercortisolaemia.

Etomidate (R 16659) is a potent GABAA receptor agonist. Etomidate is a neurological drug and effective parenteral medication and has the potential for management of endogenous hypercortisolaemia.

Specifications

Chemical Name or Material Etomidate
CAS 33125-97-2
Color White
Purity Grade Notes Research
Molecular Formula C14H16N2O2
Quantity 10 mg
Synonym R 16659
Solubility Information DMSO : ≥ 100 mg/mL (409.35 mM)
SMILES O=C(C1=CN=CN1[C@@H](C2=CC=CC=C2)C)OCC
Molecular Weight (g/mol) 244.29
Formula Weight 244.29
Percent Purity 98.78%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.