missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Etomidate
GREENER_CHOICE

Product Code. 30001258
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001258 50 mg 50mg
18044072 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001258 Supplier Tocris Bioscience™ Supplier No. 1471/50
  • ONLINE EXCLUSIVE PRICE
    866.00 €
    819.00 € / 50mg
    Save 47.00 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

GABA-mimetic; selectively interacts with β2- and β3-subunit containing GABAA receptors

General anesthetic with GABA modulatory and GABA-mimetic actions; selectively interacts with β2- and β3-subunit containing GABAA receptors. Short acting and potent hypnotic, with low toxicity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33125-97-2
Molecular Formula C14H16N2O2

Specifications

Quantity 50 mg
Formula Weight 244.29
Purity >99%
Molecular Weight (g/mol) 244.294
Chemical Name or Material Etomidate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.