missing translation for 'onlineSavingsMsg'
Learn More

Etomidate, TRC

Product Code. 30138455
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138455 10 mg 10mg
30135053 50 mg 50mg
17902605 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138455 Supplier Toronto Research Chemicals Supplier No. TRCE93330010MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33125-97-2
Molecular Formula C14 H16 N2 O2
Molecular Weight (g/mol) 244.29
Synonym 1H-Imidazole-5-carboxylic acid, 1-[(1R)-1-phenylethyl]-, ethyl ester, 1H-Imidazole-5-carboxylic acid, 1-(1-phenylethyl)-, ethyl ester, (R)-, Imidazole-5-carboxylic acid, 1-(α-methylbenzyl)-, ethyl ester, (R)-(+)-, (+)-Etomidate, Amidate, Amidate (pharmaceutical), D-Etomidate, Etomidate, Hypnomidate, Lipuro, Propiscin, R 16659, Radenarcon, d-Etomidate
IUPAC Name ethyl 3-[(1R)-1-phenylethyl]imidazole-4-carboxylate
SMILES CCOC(=O)c1cncn1[C@H](C)c2ccccc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H16N2O2/c1-3-18-14(17)13-9-15-10-16(13)11(2)12-7-5-4-6-8-12/h4-11H,3H2,1-2H3/t11-/m1/s1
Formula Weight 244.1212
Percent Purity >95%
Chemical Name or Material Etomidate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.