Learn More
R&D Systems™ Etoposide
Product Code. 18788486
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 18788486
Brand: R&D Systems™ 468409606
R&D Systems™ Etoposide complexes with topoisomerase II and DNA to enhance doublestrand and single-strand cleavage of DNA and reversibly inhibit religation.
  • Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
  • Do not use past expiration date.
  • Blocks the cell cycle in S-phase and G2-phase; induces apoptosis in normal and tumor cell lines; inhibits synthesis of the oncoprotein Mdm2 and induces apoptosis in tumor lines that overexpress Mdm2.
  • Etoposide is provided in DMSO. The final reagent concentration is 10mM.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 μL
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.