missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Etoposide

Product Code. 16411686
Encompass_Preferred
Change view
Click to view available options
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16411686 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16411686 undefined MedChemExpress missing translation for 'supplierNo' HY13629

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Etoposide (VP-16; VP-16-213) is an anti-cancer chemotherapy agent. Etoposide inhibits topoisomerase II, thus stopping DNA replication. Etoposide induces cell cycle arrest, apoptosis and autophagy.

Specifications

Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 588.56 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 33419-42-0
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 39 mg/mL (66.26 mM)
Synonym VP-16∣ VP-16-213
Target Topoisomerase∣Autophagy∣Mitophagy∣Bacterial∣Apoptosis∣Antibiotic
Quantity 500 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.