missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Two pieces vinyl tubing, Two complete male-female Quickfit connectors and cassette removal tool.
- Runs up to six lab-cast or precast gels (26 × 20cm) simultaneously
- Used with 18- or 24cm Immobiline DryStrip gels
- Simple-to-use 1.0 or 1.5mm pre-assembled hinged gel cassettes
- Innovative modular design simplifies loading and unloading
- Built-in ceramic heat exchanger and buffer circulation pump ensures efficient cooling
- Improved upper buffer chamber
- Contains: Ettan DALTsix electrophoresis unit, gel caster and gradient maker, an EPS 601 power supply, and MultiTemp III thermostatic circulator
- Electrical requirements: 115V 60Hz
Compliance
The product must be used in the same state as it was delivered and connected only to other CE labeled Cytiva modules or other products as recommended. For regulatory details please see declaration of conformity.
Specifications
Specifications
| Certifications/Compliance | CE |
| Running Time | 4 to 6.5 hr. |
| Includes | Two pieces Vinyl tubing, two complete male female QuickFit™ connectors and cassette removal tool, Ettan Daltsix electrophoresis unit, gel caster and gradient maker, an Eps 601 power supply and MultiTemp III thermostatic circulator |
| No. of Gels | 1 to 6 |
| Temperature (English) Operating | 39.2°F to 104°F |
| Temperature (Metric) Operating | 4°C to 40°C |
| Volume (English) Buffer | 152.16 oz. |
| Volume (Metric) Buffer | 4500 mL |
| Voltage | 600 V |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?