missing translation for 'onlineSavingsMsg'
Learn More

Eugenol, TRC

Product Code. 30136616
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136616 100 g 100g
30132755 10 g 10g
17942905 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136616 Supplier Toronto Research Chemicals Supplier No. TRCE938640100G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10 H12 O2
Molecular Weight (g/mol) 164.2
Synonym Eugenol, 2-Methoxy-4-(prop-2-enyl)phenol, Phenol, 2-methoxy-4-(2-propen-1-yl)-, Phenol, 2-methoxy-4-(2-propenyl)- (9CI), Phenol, 4-allyl-2-methoxy- (8CI), 1-Allyl-4-hydroxy-3-methoxybenzene, 2-Hydroxy-5-allylanisole, 2-Methoxy-1-hydroxy-4-allylbenzene, 2-Methoxy-4-(2-propen-1-yl)phenol, 2-Methoxy-4-(2-propenyl)phenol, 2-Methoxy-4-(2'-propenyl)phenol, 2-Methoxy-4-[2-allyl]phenol, 2-Methoxy-4-allylphenol, 3-(3-Methoxy-4-hydroxyphenyl)propene, 3-(4-Hydroxy-3-methoxyphenyl)-1-propene, 4-Allenylguaiacol, 4-Allyl-1-hydroxy-2-methoxybenzene, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol, 4-Hydroxy-3-methoxyallylbenzene, AQUI-S 20E, Allylguaiacol, Bioxeda, Caryophyllic acid, Dentogum, Eugenic acid, NSC 209525, NSC 8895, p-Allylguaiacol, p-Eugenol
IUPAC Name 2-methoxy-4-prop-2-enylphenol
SMILES COc1cc(CC=C)ccc1O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3,5-7,11H,1,4H2,2H3
Formula Weight 164.0837
Chemical Name or Material Eugenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.