missing translation for 'onlineSavingsMsg'
Learn More

Eumulgin 62 (Technical Grade), TRC

Product Code. 30138249
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138249 100 g 100g
30008597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138249 Supplier Toronto Research Chemicals Supplier No. TRCE525350100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68439-49-6
Synonym Macrogol 20 Cetostearyl Ether, Alcohols, C16-18, ethoxylated, Cetomacrogol 1000, A 6, AT 11, Alfonic 1618-46, Alfonic 1618-60, Alfonic 1618-62, Alfonic 1618-78, Alfonic 1618-80, B 2050-01A, Berol 07, Brij 68, Brij CS 20, C16-18 alcohols, ethoxylated, C16-18 ethoxylated alcohols, C16-18 ethoxylated alcs;C16-18 fatty alcs., ethoxylated, CE 50, Cemulsol DB, Cetomacrogol, Cetomacrogol 1000BP, Cetostearyl alcohol ethoxylated, Cremophor A 25, Cresmer 1000, Disponil O 5, Disponil OC 5, Disponil TA 11, Disponil TA 14, Empilan KLA 6, Emuldac AS 11, Emuldac AS 25, Emuldac AS 80, Ethal 368, Ethal CSA 10, Ethal CSA 17, Ethal CSA 40/70, Ethoxylated C16-18 alcohol, Ethoxylated C16-18 alcs.;Ethoxylated C16-18 fatty alcohols, Ethoxylated alcohols, C16-18, Ethoxylated cetostearyl alcohol, Ethoxylated cetostearyl alcohols, Ethoxylated cetyl stearyl alcohol, Ethylan 68-30;Eumulgin 62, Findet 1618A18, Findet 1618A23, Genapol T 250P, Hetoxol CS 20, Hetoxol CS 25, Hetoxol CS 4, Imbentin 168S300, Incropol CS 20, KM 11, KM 20, Kolliphor CS 12, Kolliphor CS 20, Laffonics CM 100, Lipal 15CSA, Lipocol SC 10, Lipocol SC 4, Lubrol W1, Lutensol AT, Lutensol AT 100, Macol CSA, 10, Macol CSA 20, Macol CSA 40, Macrogol 20 cetostearyl ether, Macrogol 260 cetylstearyl ether, Macrogol cetostearyl ether, Marlipal 1618/10, Marlowet TA 6, Mergatil CS 33A, Mergital CS 15, Mergital CS 25, Mergital CS 50A, Mergital E 1471, Neonol P 1618-10, Neonol P 1618-11, Neonol P 1618-12, OTsS 21, Oxanol TsS 21;Pareth 11, Peregal O 15, Peregal O 30, Peregal O 5, Pingpingjia O 30, Plurafac A 38, Plurafac A 39, Plurafac LF 7000, Polyoxyl 20 cetostearyl Ether, Praewozell F 1618/5;Prevocell F 1618/5, Procol CS 20, RC 100, Rhenosin RC 100, Rhodasurf ISA 2, Rokanol OL 1, Rovol T 500, Siligen WL, Simulsol CS, Sinnovax NEVA;Sinnowax AO, Sinopol 1830, Siponic E 0, Slovasol 6811, Surfacare T 20, Surfonic L 68-20, Syntanol TsS, Teginacid C, Teric 16A, Ultrol CE 200, Ultrol CE 200F, Volpo CS;

Specifications

Quantity 100 g
Chemical Name or Material Eumulgin 62
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.