missing translation for 'onlineSavingsMsg'
Learn More

Eupatorin, TRC

Product Code. 30387100
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387100 5 mg
30377433 10 mg
30374279 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387100 Supplier Toronto Research Chemicals Supplier No. TRCE9388405MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 855-96-9
Molecular Formula C18 H16 O7
Molecular Weight (g/mol) 344.32
Synonym 3',5-Dihydroxy-4',6,7-trimethoxyflavone, 5,3'-Dihydroxy-6,7,4'-trimethoxyflavone, 6-Methoxyluteolin 4',7-dimethyl ether, Eupatorin, Eupatorine, NSC 106402, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxy-4H-1-benzopyran-4-one
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxychromen-4-one
SMILES COc1ccc(cc1O)C2=CC(=O)c3c(O)c(OC)c(OC)cc3O2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H16O7/c1-22-12-5-4-9(6-10(12)19)13-7-11(20)16-14(25-13)8-15(23-2)18(24-3)17(16)21/h4-8,19,21H,1-3H3
Formula Weight 344.0896
Chemical Name or Material Eupatorin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.