missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) carbonate hydrate, REacton™, 99.99% (REO)

Product Code. 11316668
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11316668 1 g 1g
11306668 5 g 5g
11326668 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11316668 Supplier Thermo Scientific Alfa Aesar Supplier No. 013212.03
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 86546-99-8 | C3Eu2O9 | 483.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86546-99-8
Molecular Formula C3Eu2O9
Molecular Weight (g/mol) 483.95
MDL Number MFCD00149702
InChI Key MBFNJTXDYDGXDL-UHFFFAOYSA-H
Synonym europium iii carbonate hydrate, 3co3.2eu.h2o, europium iii carbonate hydrate, reacton, dieuropium 3+ hydrate tricarbonate
PubChem CID 91886282
IUPAC Name dieuropium(3+) tricarbonate
SMILES [Eu+3].[Eu+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99% (REO)
Formula Weight 483.95 (Anhydrous)
Odor Odorless
Appearance White powder
Chemical Name or Material Europium(III) carbonate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.