missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) chloride hexahydrate, 99.99%, (trace metal basis)

Product Code. 10529633 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10529633 25 g Glass bottle 25g
10786233 1 g Glass bottle 1g
10625082 5 g Glass bottle 5g
3 options
This item is not returnable. View return policy

Product Code. 10529633

Brand: Thermo Scientific Acros 193010250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13759-92-7 | Cl3Eu·6H2O | 366.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13759-92-7
Molecular Formula Cl3Eu·6H2O
Molecular Weight (g/mol) 366.4
MDL Number MFCD00149701

Specifications

Melting Point 850.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 25 g
Assay Percent Range 39.0 to 42.5% (Eu) (Complexometry)
Linear Formula EuCl3·6H2O
Fieser 15,186
Merck Index 10, 3854
Solubility Information Solubility in water: soluble
Formula Weight 366.4
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 4.89
Density 4.8900g/mL
Chemical Name or Material Europium(III) chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.