missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxide, 99.99%, (trace metal basis)

Product Code. 10513371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10513371 1 g Glass bottle 1g
10635082 5 g Glass bottle 5g
10056333 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10513371 Supplier Thermo Scientific Acros Supplier No. 193020010
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
PubChem CID 159371
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Color White to Pink
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.99%
Linear Formula Eu2O3
Merck Index 10, 3854
Solubility Information Solubility in water: insoluble
IUPAC Name dieuropium(3+) trioxidandiide
Formula Weight 351.91
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Specific Gravity 7.42
Density 7.4200g/mL
Chemical Name or Material Europium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.