missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ CyberScan pH 600 Meter

Product Code. 12951688
Change view
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) Unit Size
12951688 -2 to 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12951688 Supplier Thermo Scientific™ Supplier No. 01X415102
  • ONLINE EXCLUSIVE PRICE
    840.00 €
    774.00 € / Pack of 1
    Save 66.00 €
    7% Off
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ waterproof CyberScan pH 600 Meter is an ideal choice for routine applications in laboratories, productions plants and schools.

Features:

  • Higher accuracy under varying temperature with Automatic/ Manual Temperature Compensation.
  • Mega-memory space that stores/ recalls up to 500 Data Sets with temperature readings, with time and date-stamping formatted to meet GLP standards.
  • Wireless and cable-free data transfer with infrared IrDA technology. Comes with PC-based Complimentary Data Acquisition Software for hassle-free data import and report-generation into any popular spreadsheet programs of your choice.
  • Recall Electrode Slope, Electrode Offset Value and Previous Calibration Values for easy reference
  • Large, Easy-to-Read Custom Display with simultaneous readings provides optimum viewing with user-friendly icons
  • Hold Function momentarily freezes reading for easy viewing and recording
  • Self-Diagnostic with message codes for easy troubleshooting
  • Volatile Memory holds settings and stored data even when you run out of batteries.
  • pH Buffer Options: USA, NIST, DIN, PWB

Applications

  • Industrial: For checks in water conditioning plants, conditioning plants, cooling towers, food processing water testing, printing and chemical industries, and water/wastewater treatment.
  • Environmental/Agricultural: Use in ecology studies, aquariums, and hydroponics.
  • Educational: Ideal for quick, accurate routine pH checks in laboratories and schools.
TRUSTED_SUSTAINABILITY

Specifications

Description pH600 pH/mV Meter with ATC Probe & DAS software
Includes Stability Indicator; Self-diagnostic; Non-Volatile Memory; Hold & Auto-Power Off after 20 minutes
Accuracy (Temperature) 0.1°C/1.0°F
Accuracy (mV) ±0.2
Accuracy (pH) ±0.01
Outputs IrDA, RS-232C
Measurement Modes 500
Calibration Custom Buffer Calibration
Range (pH) -2 to 20
Resolution (pH) 0.01, 0.1
Range (mV) ±2000
Resolution (mV) 0.1 mV
Calibration Points 2 to 5
Temperature Range (Metric) -10°C to +110°C
Temperature Range (English) 14.0°F to 230°F
Resolution (Temperature) 0.1°C/1.0°F
Display Type LCD, Backlit Dot Matrix, Slope/ Offset Display: Yes (Display + Icon)
Memory 500 data sets
Data Points 500
Alarm Output Cal Due Alarm: Yes (Days - Max 30), Set-Point Alarm
Inputs DC phono sockets, 8 pin connector, BNC
Certifications/Compliance GLP: Yes
IP Rating IP67
AC Adapter 9VDC power adapter (optional)
Power Supply 9VDC power adapter
Temperature Compensation Automatic/Manual (0°C to 100°C)
Battery Type 4 AAA x 1.4V batteries
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.