missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ EC620131 pH electrode

Product Code. 12624830
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Unit Size
12624830 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12624830 Supplier Thermo Scientific™ Supplier No. 01X218973
  • ONLINE EXCLUSIVE PRICE
    216.08 €
    193.00 € / Pack of 1
    Save 23.08 €
    10% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Features:

  • Buffers and standard solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Description Open pore glass-body pH electrode for general testing and high viscosity solutions where frequent cleaning of reference is required
Connector Types BNC
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 80°C
Type pH Electrode
For Use With (Application) pH measurement
For Use With (Equipment) All pH meters with BNC input connector
Temperature (Metric) Range 0°C to 80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.