missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Ion 6+ meter

Product Code. 11549174
Change view
Click to view available options
Includes:
ATC Electrode
pH Electrode
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
11549174 ATC Electrode Each
11559174 pH Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11549174 Supplier Thermo Scientific™ Supplier No. 01X256409
  • ONLINE EXCLUSIVE PRICE
    535.00 €
    508.00 € / Pack of 1
    Save 27.00 €
    5% Off
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Quick and easy measurement with this ion-selective meter.

Quick and easy measurement with this ion-selective meter. Applications for the Eutech™ Ion 6+ meter include: ISE (Use with any ISE with BNC connection), water quality testing, environmental/agricultural testing, laboratories and schools, and in industrial settings.

Features

  • Direct Ion Concentration for use with most mono- or di-valent Ion Selective Electrodes (ISE)
  • mV measurement mode allows you to check performance and calibration of your ISE
  • Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Non-Volatile Memory ensures data are retained even meter powers off
  • Hold Function momentarily freezes reading for easy viewing and recording
  • Auto-Power Off after 17 minutes saves battery power after non-use
  • Self-Diagnostic with message codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
  • pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA), 1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (ISE) ± 1% full scale
Accuracy (mV) 1 mV & ±2 mV (beyond ±200 mV)
Accuracy (pH) ± 0.01
Accuracy (Temperature) ±0.5°C
Battery Type 4 AAA x 1.4V
Calibration Points (pH) Up to 5 points
Description Eutech Ion 6+ Ion/pH/mV Meter with ATC Electrode ECPHTEM01P, Rubber Armour/Stand
Calibration Points (ISE) Up to 5
Measurement Modes Ion-Selective Meter
Range (ISE) Ion Concentration: 0.01 to 0.99 ppm, 1.0 to 199.9 ppm, 200 to 1999 ppm
Range (mV) ±500 mV
Range (pH) 0 to 14
Resolution (ISE) 0.01 ppm, 0.1 ppm, 1 ppm
Resolution (pH) 0.01
Temperature (Metric) Range 0°C to 100.0°C
Includes ATC Electrode
Resolution (mV) 0.1 mV and ±0.2 mV (within ±199.9 mV)
Resolution (Temperature) 0.1°C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.