missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ PC 2700 Multi-parameter Meter

Product Code. 11523865
Change view
Click to view available options
Product Type:
Meter with ph Electrode
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11523865 Meter with ph Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11523865 Supplier Thermo Scientific™ Supplier No. 01X543906
  • ONLINE EXCLUSIVE PRICE
    1610.00 €
    1530.00 € / Pack of 1
    Save 80.00 €
    4% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.

Your all-in-one solution for multi-parameter research work in the lab. The Eutech™ PC 2700 Multi-parameter Meter is accurate, reliable and intuitive with user-friendly functions. Meter comes with a multi-data screen that includes temperature, electrode status, calibration points, date and time along with your desired measurement parameter. PC 2700 measures pH, ORP, ION, Conductivity, TDS, Salinity, or Resistivity. Configuration include 30X427301 RS232 cable, 100/240 VAC power adapter and an integral electrode holder.

Features

  • Choose auto-calibration with preset values for quick easy calibration, ormanualmulti-point calibration for greater accuracy
  • Easy standardization with auto-standardization feature – detect the exact cell constant value of your electrodes with the press of a button
  • Up to 6-point calibration with auto-buffer recognition
  • Includes high performance 4-cell Conductivity probe
  • Quick, easy electrode diagnosis with the effective cell constants display
  • Non-volatilememory holds up to 500 data points – time and date-stamped for GLP compliance
  • Bi-directional RS232 for easy data transfer to computer
  • Cal-due alarm – nomore out-dated calibrations!
  • Auto-logging function for convenient continuousmonitoring
  • Limit alarm alerts when reading falls out of range
  • Password protection for setup and calibration

Applications

  • Water quality testing
  • Checkingmetal finishes
  • Solutions
  • Cooling tower water
  • Printing fountain solutions
  • Boiler water
  • Brines
  • Drillingmud
  • Rinse tanks
  • Ponds
  • Pollution control
  • Recirculating systems
  • Waste water
  • Industrial process systems
  • General use in laboratories
  • Titration
  • Quality assurance
  • Ecological studies
  • Food-processing

Specifications

Product Specification: Description

pH

  • Range: -2.000 to 20.000 pH
  • Resolution: 0.1 / 0.01 / 0.001 pH
  • Accuracy: ±0.002 pH + 1 LSD
  • Cal. Points: Up to 6 preset or 5 custom
  • Buffer Sets: USA, NIST, DIN, User 1, User 2, Custom

ORP

  • Range: ±2000.0mV
  • Rel.mV Range: ±2000.0mV
  • Resolution: 0.1mV
  • Accuracy: ±0.2mV

ION

  • Concentration: 0.001 to 19999 ppm (±2000mV)
  • Resolution: 2 or 3 digits
  • Accuracy: 0.5 % full scale (monovalent) ; 1 % full scale (divalent)
  • Cal. Points: 2 to 8

Conductivity

  • Range: 0.050 μS to 500.0mS
  • Resolution: 0.01 / 0.1 μS ; 0.001 / 0.01 / 0.1mS
  • Accuracy: ±1 % full scale
  • Cal. Points: Up to 5
  • Cell Constant: 0.010 to 10.000

TDS

  • Range (Depending On TDS Factor): 0.050 ppm to 500 ppt (@ TDS factor 1.00)
  • Resolution: 0.01 / 0.1 ppm ; 0.001 / 0.01 / 0.1 ppt
  • Accuracy: ±1 % full scale
  • Cal. Points: Up to 5

Salinity

  • Range: 0.0 to 80.0 ppt
  • Resolution: 0.01 / 0.1 ppm ; 0.001 / 0.01 / 0.1 ppt
  • Accuracy: ±1 % full scale

Resistivity

  • Range: 2.000 Ohm to 20.0mOhm
  • Resolution: 0.01 / 0.1 Ohm ; 0.001 / 0.1 kOhm ; 0.01mOhm
  • Accuracy: ±1 % full scale

Temperature

  • Range (meter): 0.0 to 100.0 °C / 32.0 to 212.0 °F
  • Resolution: 0.1 °C / 0.1 °F
  • Accuracy: ±0.3 °C / ±0.5 °F
  • Coefficient (Per ?C): Linear & pure ; 0.000 to 10.000 %
  • Normalization: 15.0 to 30.0 °C / 59.0 to 86.0 °F
  • Compensation: ATC /mTC
  • Power Requirements: 9 V DC adapter, 1.3 A (100/240 VAC, SmPS)
TRUSTED_SUSTAINABILITY

Specifications

Outputs RS232
Description Eutech PC 2700 Meter With pH Electrode (ECFG73711B), Conductivity Electrode (CONSEN921D), ATC Probe (PH5TEMB1P), Integral Electrode Holder & 1/24 VAC Adapter, RS232 cable, & pH Electrode Refill Solution
Measurement Modes pH, Conductivity
Range (Conductivity) 0.050 μS to 500 mS
Range (pH) -2 to 20
Range (Resistivity) 2 Ω to 20 mΩ
Range (Salinity) 0 to 80 ppt
Range (TDS) 0.01 ppm, 0.1 ppm, 0.001 ppt, 0.01 ppt, 0.1 ppt
Resolution (pH) 0.1, 0.01, 0.001
Temperature Range (Metric) 0°C to 100°C
Product Type Meter with ph Electrode
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.