missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ PC 450 Meter with pH /Con/ATC Electrodes

Product Code. 15337063
Change view
Click to view available options
Measurement Modes:
pH, Conductivity, ATC Electrodes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Modes Unit Size
15337063 pH, Conductivity, ATC Electrodes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15337063 Supplier Thermo Scientific™ Supplier No. 01X651639
  • ONLINE EXCLUSIVE PRICE
    1095.00 €
    1040.00 € / Pack of 1
    Save 55.00 €
    5% Off
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Eutech™ offers a wide range of pH meters and electrodes. pH 450 Meter is a rugged waterproof meter ideal for any environment and offers measurement of pH / mV / Temperature.

Features

  • Durable waterproof and dustproof design with IP67 rating
  • Large, backlit display – Viewmeasurement with temperature and % slope
  • Grip-Clip™ – Quickly and easily secures the pH electrode to your beaker or container
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). Newmanual option allows calibration with up to 5 custom buffer standards
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate themeter
  • Ready indicator,manual or automaticmeasurement hold feature and optional automatic shutoff function
  • Selectable °C/°F – Choose units to best fit your application
  • Intuitive, user-friendly icons – Electrode status displays pH slope
  • Store up to 500 data sets – Easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date –meets standards for GLP (Good Laboratory Practices)
  • Extended battery life from two AA batteries, or use optional universal power adapter
  • Built-in stand with wall-mount option – Allows for easy benchtopmeasurement or storage
  • Compatible with pH, ORP or ISE electrodes having a BNC connector

Specifications

Product Specification: Description

pH

  • Range: -2.00 to 16.00 pH
  • Resolution: 0.01 pH
  • Accuracy: ±0.01 pH
  • Calibration: Up to 5 points (USA & NIST & CUSTOm), Up to 6 points (DIN)

mV

  • Range: ±2000mV
  • Resolution: 0.1mV ±999.9mV; 1mV beyond
  • Accuracy: ±0.2mV +2 LSD or +0.5 % of reading; whichever is greater
  • Calibration: ±200mV

ION

  • Range: 0.0 to 2000 ppm
  • Resolution: 0.01 / 0.1 / 1 ppm
  • Accuracy: ±0.5 % full scale (monovalent); ±1 % full scale (divalent)
  • Calibration: From 2-4 or 2-5 points: 0.10, 1.00, 10.0, 100.0 & 1000 / 0.20, 2.00, 20.0, 200.0 & 2000 / 0.50, 5.00, 50.0 & 500.0

Temperature

  • Range: -10.0 to 110.0 °C (14.0 to 230.0 °F)
  • Resolution: 0.1 °C/°F
  • Accuracy: ±0.5 °C; ±0.9 °F
  • Calibration: Offset in 0.1 increments, ±10 °C or ±18 °F

Others

  • Display: Custom LCD with backlight
  • memory: Stores up to 500 data sets
  • Output: USB or RS232
  • Real-Time Clock: Time and date stamp on calibration and stored data
  • Temperature Compensation: Automatic ormanual from -10 to 110 °C
  • Power: Two AA batteries (included), >500 hours (without backlight); AC adapter (optional)
  • Dimensions (L x W x H):meter: 7 7/8 x 3 1/8 x 2 1/4 in. (20 x 8.3 x 5.7 cm), Boxed: 9 1/8 x 9 1/8 x 2 3/4 in. (23 x 23 x 7 cm)
  • Weight:meter: 1 lb (0.5 kg), Boxed: 1.4 lb (0.7 kg)
TRUSTED_SUSTAINABILITY

Specifications

Outputs USB, RS232
Description PC 450 Meter & pH & Con/ATC Electrodes: Meter, double-junction pH electrode, conductivity cell (K = 1.0), Grip-Clip™, two AA batteries
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, Conductivity, ATC Electrodes
Resolution (pH) 0.01
Temperature Range (Metric) -10°C to +110°C
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.