missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ PC 450 Multi-parameter Meter Kit

Product Code. 15367063
Change view
Click to view available options
Measurement Modes:
pH, ORP, Ion, Conductivity, TDS, Salinity, Resistivity
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Modes Unit Size
15367063 pH, ORP, Ion, Conductivity, TDS, Salinity, Resistivity Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15367063 Supplier Thermo Scientific™ Supplier No. 01X651638
  • ONLINE EXCLUSIVE PRICE
    1180.00 €
    1120.00 € / Pack of 1
    Save 60.00 €
    5% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.

Your all-in-one solution for multi-parameter research work in the lab. The multi-parameter Eutech PC 450 measures pH / mV / Conductivity / Total Dissolved Solids / Temperature. It accepts a single pH/Con probe or separate pH and conductivity probes.

Features

  • Durable waterproof and dustproof design with IP67 rating
  • Switch between pH, mV, conductivity and TDS with a press of a button
  • Large, backlit custom display – Lets you view measurement with temperature
  • Grip-Clip™ – Quickly and easily secures separate pH and conductivity electrodes simultaneously to your sample container or beaker
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards you
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
  • Intuitive, user-friendly icons – Electrode status displays pH slope or cell constant
  • Auto-range conductivity from 0 to 199.9 mS in five ranges – With up to five calibration points
  • Selectable cell constant – 0.10, 1.00 or 10.0
  • Adjustable temperature coefficient – From 0.00 to 10.00 % per ºC
  • Conductivity-to-TDS factor – Adjustable from 0.40 to 1.00
  • Selectable ºC/ºF – Choose units to best fit your application
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • Store up to 500 data sets; easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
  • 500 hours battery life or use optional universal power adapter
  • Built-in stand with wall-mount – Allows for easy benchtop measurement and storage
TRUSTED_SUSTAINABILITY

Specifications

Outputs USB or RS232
Accuracy (Conductivity) ± 1% full scale
Accuracy (mV) ±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater
Accuracy (pH) ±0.01
Accuracy (TDS) ± 1% full scale
Calibration Points (Conductivity) Auto/manual; one point per range
Calibration Points (pH) Up to 6 points: USA, NIST, DIN or custom
Compensation Temperature: Automatic or manual from -10 to 110 °C
Description PC 450 Meter Combi Kit: Meter, combi pH/Con/ATC electrode, two AA batteries, hard carry case, USB cable, calibration solutions
Display Type LCD Graphic, Backlit
IP Rating IP67
Measurement Modes pH, ORP, Ion, Conductivity, TDS, Salinity, Resistivity
Range (Conductivity) 0 to 200 mS, 0 to 20 mS if using COMBI03BW probe
Range (mV) ±2000 mV
Range (pH) -2 to 16
Range (TDS) 0 to 200 ppt (TDS factor 1.0)
Resolution (pH) 0.01
Temperature Range (Metric) -10.0°C to +110.0°C
Memory Stores up to 500 data sets
Power Supply Two AA batteries (included), >100 hours (without backlight); AC adapter (optional)
Resolution (Conductivity) 0.01 μS to 0.1 mS
Resolution (mV) 0.1 mV ±999.9 mV; 1 mV beyond
Resolution (TDS) 0.01 ppm to 0.1 ppt
Temperature Accuracy ±0.5°C, ±0.9°F
Temperature Range (English) 14.0°F to 230.0°F
Temperature Resolution 0.1 °C/°F
Dimensions (L x W x H) Meter: 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm)
Weight (English) Meter: 1 lb
Weight (Metric) Meter: 0.5 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.