missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ PD 650 Meter

Product Code. 10233473
Change view
Click to view available options
Range (pH):
-2 to 20
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
10233473 -2 to 20 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10233473 Supplier Thermo Scientific™ Supplier No. 01X430702
  • ONLINE EXCLUSIVE PRICE
    2305.00 €
    2130.00 € / Pack of 1
    Save 175.00 €
    7% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Collecting multi-parameter data is a breeze using Eutech™ PD 650 Meter.

Eutech™ PD 650 Meter is a multi-paramater meter that measures pH / ORP / Ion / Dissolved Oxygen / Temperature. Collecting multi-parameter data in the field is a breeze with Eutech's CyberScan 650 multi-parameter series. The CyberScan PD650 delivers quick, lab-accurate measurements of pH and dissolved oxygen simultaneously. Auto-logging function records up to 500 data sets with date/time in GLP-compliant format. A comprehensive screen displays pH, dissolved oxygen and temperature readings simultaneously in addition to electrode status reports and calibration information.

Features:

  • ±1% full scale accuracy at 3-decimal resolution across a wide measurement range
    Ion-measurement capabilities with 3-digit resolution
  • Selectable multi-point calibration for more accurate conductivity measurements or single-point calibration for quick, convenient calibration with no mistakes
  • Measures 90.00mg/L in DO concentration and 600% in DO saturation
  • Fast and easy DO calibration with 100% calibration option against atmospheric air or 0% calibration against a zero solution
  • Salinity, Temperature and Pressure Compensation — accurate readings under varying conditions
  • No warm up time required with galvanic system of measurement
  • One-Glance-Tells-All Screen Display with Backlight Electrode status report, calibration data and simultaneous display of up to 4 parameters
  • Easy-to-understand prompt messages for easy set-up, calibration and trouble-shooting
  • Recorded readings individually stamped with time and date in GLP compliant format
  • Auto-logging meter saves readings automatically within seconds to prevent data loss
  • Wireless Information Technology: No wires, no cables—Transfer data from meter to computer with the press of a button
  • Non-Volatile Memory protects information and meter settings, even when batteries run out
  • Four Levels of Password Protection prevents tampering or accidental data deletion
  • pH Buffer Options: USA, NIST, DIN, PWB, Custom
TRUSTED_SUSTAINABILITY

Specifications

Outputs IrDA Wireless Communications
Accuracy (Barometer) 0.1 mmHg or 0.1 kPa
Accuracy (DO) ±0.2mg/L / ±0.2%
Accuracy (ISE) ±0.5% FS (mono); 1% FS (div)
Accuracy (mV) ±0.2 mV
Accuracy (pH) ±0.002
Accuracy (Salinity) 0.1 ppt
Alarm Output Calibration Due Alarm, HI-LO limit (out of range reading) Alarm
Barometric Pressure Correction Automatic correction with built-in sensor
Battery Type 4 X 1.5 V AA Alkaline
Calibration Points (ISE) Up to 8
Calibration Points (pH) 1 (Offset) to 6 points
Description Waterproof CyberScan PD 650. pH/ mV/ Ion/ Dissolved Oxygen, handheld meter with pH electrode ECFC252203B, Dissolved Oxygen probe (DOHANDYNEW), DAS software, power adapter & carrying kit set (all probes comes with 3m long cable.)
Display Type Multi-data Backlit Dot Matrix, 7.2 cm (H) X 5.4 cm (W)
IP Rating IP67
Measurement Modes pH, ORP, Ion, Dissolved Oxygen, Temperature
Range (mV) ±2000.0 mV
Range (pH) -2 to 20
Temperature Range (Metric) -10.0°C to +110.0°C (Meter)
Includes Auto-Calibration; Data-logging options with time/date indexed, GLP compliance; Simultaneous Display and measurement of 4 parameters; Cal-due alarm; pH slope/offset Display & Self-diagnostic
Memory 500 Data Sets with Date/Time stamping & Security password protection
Percent Saturation Range (RDO) 0 to 90.00 mg/L or ppm / 0 to 600%
Power Supply 9 V DC adapter, 500 mA
Pressure Resolution 450 to 825 mmHg / 59.9 to 109.9 kP
Range DO: 0 to 90.00 mg/L or ppm / 0 to 600%
Range (ISE) 0.001 to 19900 ion concentration
Resolution DO: 0.01 mg/L or ppm/ 0.1%
Resolution (mV) 0.1 mV
Resolution (Salinity) 0.0 to 50.0ppt
Salinity Factor Salinity Correction - Automatic; Resolution & Accuracy - 0.0 to 50.0ppt & 0.1 ppt
Temperature Accuracy ±0.5°C, ±0.9°F
Temperature Compensation Automatic / Manual (0 to 100 °C)
Temperature Range (English) 14.0°F to 230.0°F (Meter)
Temperature Resolution 0.1 °C/°F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.