missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ PD450 Meter Kit ECPDWP45004K

Product Code. 15337073
Change view
Click to view available options
Measurement Modes:
pH, mV, Dissolved Oxygen, Temperature
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Modes unitSize
15337073 pH, mV, Dissolved Oxygen, Temperature Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15337073 Supplier Thermo Scientific™ Supplier No. 01X651642
  • ONLINE EXCLUSIVE PRICE
    1875.00 €
    1780.00 € / Pack of 1
    Save 95.00 €
    5% Off
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Collecting multi-parameter data is a breeze using Eutech™ PD450 Meter Kit ECPDWP45004K.

The Eutech™ PD450 Meter is a multi-parameter meter that measures pH, mV, dissolved oxygen, and temperature. Affordable optical RDO technology reduces maintenance time and costs. The maintenance free optical probe with no electrolyte eliminates the hassle of replacing delicate membranes and salt build up from refillling solutions. You can also easily switch between pH, mV, % saturation and mg/L (ppm) dissolved oxygen – with the press of a button.

Features:

  • pH / mV / Dissolved Oxygen / Temperature
  • Durable waterproof and dustproof design with IP67 rating
  • Large, backlit graphic display provides measurement with temperature
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
  • Selectable ºC or ºF – Choose units to best fit your application
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • Store up to 500 data sets – Easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
TRUSTED_SUSTAINABILITY

Specifications

AC Adapter Optional
Outputs USB or RS232
Accuracy (DO) ±2 %; ±0.1 ppm or mg/L up to 8; ±0.2 ppm or mg/L above 8
Accuracy (mV) ±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater
Accuracy (pH) ±0.01
Battery Life 100+ hr. (without backlight)
Battery Type 2 AA (included)
Calibration mV: ±200 mV, DO: Zero and saturation, Temp: Offset in 0.1 increments, ±10 °C or &
Calibration Points (pH) Up to 6 points: USA, NIST, DIN or custom
Compensation Pressure & Salinity Compensation: manual
Description PD 450 Meter Kit: Meter, pH electrode, RDO electrode with sensor cap, two AA batteries, hard carry case, USB cable, RDO stainless steel weight/guard, RDO calibration chamber, calibration solutions
Display Type Backlit Custom LCD
IP Rating IP67
Measurement Modes pH, mV, Dissolved Oxygen, Temperature
Range (mV) ±2000 mV
Range (pH) -2 to 16
Range (Salinity) 0 to 80 ppt
Resolution (pH) 0.01
Temperature Range (Metric) -10.0°C to +110.0°C
Includes Time and date stamp on calibration and stored data
Memory Stores up to 500 data sets
Range (Polarographic DO) 0 to 200 %; 0 to 20 ppm/mg/L
Resolution (mV) 0.1 mV ±999.9 mV; 1 mV beyond
Resolution (Polarographic DO) 0.10 %; 0.1 ppm/mg/L
Temperature Accuracy ±0.5°C, ±0.9°F
Temperature Compensation Automatic or manual from -10 to 110 °C
Temperature Range (English) 14.0°F to 230.0°F
Temperature Resolution 0.1 ºC/ºF
Dimensions (L x W x H) Meter: 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm), Boxed: 9 1/8in x 9 1/8in x 2 3/4in (23 x 23 x 7 cm)
Weight (English) Meter: 1 lb , Boxed: 1.4 lb
Weight (Metric) Meter: 0.5 kg, Boxed: 0.7 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.