missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 150 Meter Kit

Artikelnummer. 15387043
Ansicht ändern
Click to view available options
Type:
pH Meter
pH Meter and Electrode
Packungsgröße:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Type Packungsgröße
15387043 pH Meter and Electrode Each
15377043 pH Meter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15387043 Lieferant Thermo Scientific™ Lieferanten-Nr. 01X651608
  • ONLINE EXCLUSIVE PRICE
    542.70 €
    500.00 € / Pack of 1
    Save 42.70 €
    7% Off
Estimated Shipment: 16-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Eutech™ offers a wide range of pH meters and electrodes. pH 150 Meter Kit is an an IP67-rated waterproof pH meter that offers measurement of pH / mV / Temperature.

Features

  • Large, three line LCD – View pH or mV measurement with temperature and % slope
  • Built-in memory function – Stores up to 150 measurements along with temperature
  • Auto buffer recognition – Automatically identifies USA, NIST or DIN buffer sets for rapid calibration
  • Power with AA batteries or use optional AC adapter for benchtop measurements
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
  • Selectable ºF/ºC – Choose units to best fit your application
  • Features mV offset – Calibrate to a Redox/ORP standard or use for pH electrode diagnostics
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • % slope and temperature displayed with each measurement
  • Built-in stand – Ideal for easy benchtop or long-term use
  • Wall-mount option – Ideal to save on bench space
  • Includes innovative Grip-Clip™ – Quickly and easily secures the electrode to your beaker or container
  • Compatible with pH electrodes having a BNC connector
TRUSTED_SUSTAINABILITY

Spezifikation

Description pH 150 Meter & Electrode: Meter with double-junction pH electrode ECFC7252201B and ATC probe PHWPTEM01W, Grip-Clip™, two AA batteries
Accuracy (Temperature) ±0.5°C;/±0.9°F
Accuracy (mV) ±0.2 or ±0.05%, whichever is greater
Accuracy (pH) ±0.01
Calibration mV: ±200 mV, Temp: Offset in 0.1 increments, ±10°C or ±18°F
Range (pH) -2 to 16
Resolution (pH) 0.01
Resolution (mV) 0.1 mV ±999.9 mV; 1 mV beyond
Calibration Points Up to 5 (USA & NIST), Up to 6 (DIN)
Temperature Range (Metric) -10°C to +110°C
Temperature Range (English) 14.0°F to 230.0°F
Resolution (Temperature) 0.1°C/°F
Display Type LCD, 3-Line Custom
Memory Stores up to 150 data sets
Data Points 150
Dimensions (L x W x H) 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm)
Weight (Metric) 0.5 kg
Weight (English) 1 lb.
Power Supply Two AA batteries (included), >700 hours continuous use; AC adapter (optional)
Range (Relative mV) ±2000
Temperature Compensation Automatic/Manual (-10°C to 110°C)
Type pH Meter and Electrode
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.