missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 2700 pH Meter

Product Code. 11573855
Change view
Click to view available options
Includes:
Intergral Electrod Holder
pH Electrode, ATC Probe
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
11573855 Intergral Electrod Holder Each
11583855 pH Electrode, ATC Probe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11573855 Supplier Thermo Scientific™ Supplier No. 01X543901
  • ONLINE EXCLUSIVE PRICE
    1095.00 €
    1040.00 € / Pack of 1
    Save 55.00 €
    5% Off
View all promotions
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Oversize screen with large fonts yet compact – the Eutech pH 2700 pH Meter offers an easy to read screen that says more. View pH or ORP readings, with temperature, electrode status, calibration points, date and time all at once!

Features

  • Up to 6-point calibration with auto-buffer recognition
  • Quick, easy electrode diagnosis with multiple pH slopes and offset display
  • Non-volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Calibrate with up to 5 custom pH buffers – use any pH values that are ≥1.0 pH unit apart
  • Cal-due alarm – no more out-dated calibrations!
  • Auto-logging function for convenient continuous monitoring
  • Password protection for setup and calibration
  • Buffer Sets: USA, NIST, DIN, User 1, User 2, Custom

Applications

  • Educational: Useful for most laboratory, ecological studies and other applications
  • Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing, where GLP data-management is required. Use in all types of food processing.
TRUSTED_SUSTAINABILITY

Specifications

Description Eutech pH 2700 Meter With Integral Electrode Holder & 100/240 VAC Adapter, RS232 cable
Includes Intergral Electrod Holder
Accuracy (Temperature) ±0.3°C/±0.5°F
Accuracy (mV) ±0.2
Accuracy (pH) ±0.002
Outputs RS-232
Range (pH) -2 to 20
Resolution (pH) 0.001, 0.01, 0.1
Range (mV) ±2000.0
Range (ORP) ±2000.0 mV
Resolution (mV) 0.1 mV
Calibration Points Up to 6 preset or 5 custom
Temperature Range (Metric) 0°C to 100°C
Temperature Range (English) 32.0°F to 212.0°F
Resolution (Temperature) 0.1°C/°F
Data Points 500 with Date and Time Stamp
Log Function Type Automatic Data Logging
Model ph 2700
Power Supply 9 V DC adapter, 1.3 A (100/240 VAC, SMPS)
Temperature Compensation ATC and MTC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.