missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ pH 450 Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
518.00 € - 616.00 €
Specifications
Accuracy (mV) | ±0.2 +2 LSD or +0.5 % of reading, whichever is greater |
---|---|
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | ±0.5°C/±0.9°F |
Outputs | USB, RS-232 |
Includes | Real-Time Clock: Time and date stamp on calibration and stored data |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15317053
|
Thermo Scientific™
01X651628 |
pH 450 Meter Only: Meter, Grip-Clip™, two AA batteries |
518.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15327053
|
Thermo Scientific™
01X651629 |
pH 450 Meter & Electrode: Meter with double junction pH electrode ECFC7252201B and ATC probe PHWPTEM01W, Grip-Clip™, two AA batteries |
616.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Eutech™ offers a wide range of pH meters and electrodes. pH 450 Meter Kit is a rugged waterproof meter ideal for any environment and offers measurement of pH / mV / Ion / Temperature.
Features
- Durable waterproof and dustproof design with IP67 rating
- Large, backlit display – View measurement with temperature and % slope
- Grip-Clip™ – Quickly and easily secures the pH electrode to your beaker or container
- Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards
- Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
- Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
- Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
- Selectable ºC/ºF – Choose units to best fit your application
- Intuitive, user-friendly icons – Electrode status displays pH slope
- Store up to 500 data sets – Easy USB or RS232 output with optional data cables
- Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
- Extended battery life from two AA batteries, or use optional universal power adapter
- Built-in stand with wall-mount option – Allows for easy benchtop measurement or storage
- Compatible with pH, ORP or ISE electrodes having a BNC connector
Specifications
±0.2 +2 LSD or +0.5 % of reading, whichever is greater | |
±0.5°C/±0.9°F | |
Real-Time Clock: Time and date stamp on calibration and stored data | |
mV: ±200 mV, Ion: From 2 to 4 or 2 to 5 points: 0.10, 1.00, 10.0, 100.0 & 1000 / 0.20, 2.00, 20.0 | |
0.01 | |
0.1 mV ±999.9 mV; 1 mV beyond | |
-10°C to +110°C | |
0.1°C/°F | |
LCD, Custom with Backlight | |
500 | |
Optional | |
Meter: 0.5 kg, Boxed: 0.7 kg | |
0.0 to 2000 ppm | |
Automatic/Manual (-10°C to 110°C) | |
>500 hr. (without backlight) |
±0.01 | |
USB, RS-232 | |
500 | |
-2 to 16 | |
±2000 | |
Up to 5 (USA & NIST & CUSTOM), Up to 6 (DIN) | |
14.0°F to 230.0°F | |
Offset in 0.1 increments, ±10 °C or ±18 °F | |
Stores up to 500 data sets | |
IP-67 | |
Meter: 7 7/8 x 3 1/8 x 2 1/4 in (20 x 8.3 x 5.7 cm), Boxed: 9 1/8 x 9 1/8 x 2 3/4 in (23 x 23 x 7 cm) | |
Meter: 1 lb., Boxed: 1.4 lb. | |
0.01 / 0.1 / 1 ppm | |
2 x AA batteries (included) | |
±0.5 % full scale (monovalent); ±1 % full scale (divalent) |