missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 450 Meter Kit

Product Code. 15337053
Change view
Click to view available options
Calibration Points:
2 to 4, 2 to 5
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibration Points Unit Size
15337053 2 to 4, 2 to 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15337053 Supplier Thermo Scientific™ Supplier No. 01X651630
  • ONLINE EXCLUSIVE PRICE
    723.60 €
    669.00 € / Pack of 1
    Save 54.60 €
    7% Off
View all promotions
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Eutech™ offers a wide range of pH meters and electrodes. pH 450 Meter Kit is a rugged waterproof meter ideal for any environment and offers measurement of pH / mV / Temperature.

Features

  • Durable waterproof and dustproof design with IP67 rating
  • Large, backlit display – Viewmeasurement with temperature and % slope
  • Grip-Clip™ – Quickly and easily secures the pH electrode to your beaker or container
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). Newmanual option allows calibration with up to 5 custom buffer standards
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate themeter
  • Ready indicator,manual or automaticmeasurement hold feature and optional automatic shutoff function
  • Selectable °C/°F – Choose units to best fit your application
  • Intuitive, user-friendly icons – Electrode status displays pH slope
  • Store up to 500 data sets – Easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date –meets standards for GLP (Good Laboratory Practices)
  • Extended battery life from two AA batteries, or use optional universal power adapter
  • Built-in stand with wall-mount option – Allows for easy benchtopmeasurement or storage
  • Compatible with pH, ORP or ISE electrodes having a BNC connector

Specifications

Product Specification: Description

pH

  • Range: -2.00 to 16.00 pH
  • Resolution: 0.01 pH
  • Accuracy: ±0.01 pH
  • Calibration: Up to 5 points (USA & NIST & CUSTOm), Up to 6 points (DIN)

mV

  • Range: ±2000mV
  • Resolution: 0.1mV ±999.9mV; 1mV beyond
  • Accuracy: ±0.2mV +2 LSD or +0.5 % of reading; whichever is greater
  • Calibration: ±200mV

ION

  • Range: 0.0 to 2000 ppm
  • Resolution: 0.01 / 0.1 / 1 ppm
  • Accuracy: ±0.5 % full scale (monovalent); ±1 % full scale (divalent)
  • Calibration: From 2-4 or 2-5 points: 0.10, 1.00, 10.0, 100.0 & 1000 / 0.20, 2.00, 20.0, 200.0 & 2000 / 0.50, 5.00, 50.0 & 500.0

Temperature

  • Range: -10.0 to 110.0 °C (14.0 to 230.0 °F)
  • Resolution: 0.1 °C/°F
  • Accuracy: ±0.5 °C; ±0.9 °F
  • Calibration: Offset in 0.1 increments, ±10 °C or ±18 °F

Others

  • Display: Custom LCD with backlight
  • memory: Stores up to 500 data sets
  • Output: USB or RS232
  • Real-Time Clock: Time and date stamp on calibration and stored data
  • Temperature Compensation: Automatic ormanual from -10 to 110 °C
  • Power: Two AA batteries (included), >500 hours (without backlight); AC adapter (optional)
  • Dimensions (L x W x H):meter: 7 7/8 x 3 1/8 x 2 1/4 in. (20 x 8.3 x 5.7 cm), Boxed: 9 1/8 x 9 1/8 x 2 3/4 in. (23 x 23 x 7 cm)
  • Weight:meter: 1 lb (0.5 kg), Boxed: 1.4 lb (0.7 kg)
TRUSTED_SUSTAINABILITY

Specifications

Description Measurement of pH / mV / Temperature
Accuracy (Temperature) 0.5°C/0.9°F
Accuracy (mV) ±0.2 or ±0.05%, whichever is greater
Accuracy (pH) ±0.01
Outputs USB, RS-232
Resolution (pH) 0.01
Calibration Points 2 to 4, 2 to 5
Temperature Range (Metric) -10°C to +110°C
Display Type LCD
Backlight Option Yes
Data Points 500
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.