Learn More
Thermo Scientific™ Eutech™ pH 700 Meter
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools.
Marke: Thermo Scientific™ 01X541605
Code : A0
Additional Details : Weight : 0.65000kg
Beschreibung
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools. It features a large, easy-to-read display, ready indicator that letsyou know when readings are stable and up to 5-point calibration with auto-buffer recognition. It also has a non-volatile memory holds up to 100 data points and comes witu an integral electrode holder.
Features
- Large, easy-to-read display
- Ready indicator let you know when readings are stable
- Up to 5-point calibration with auto-buffer recognition
- Non-volatilememory holds up to 100 data points
- Integral electrode holder
Applications
- Educational: Useful formost laboratory, ecological studies and other applications.
- Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing. Use in all types of food processing.
Specifications
Product Specification: Description
pH
- Range: -2.00 to 16.00 pH
- Resolution: 0.01 pH
- Accuracy: ±0.01 pH
- Cal. Points: Up to 5
- Buffer Sets: USA (1.68, 4.01, 7.00, 10.01, 12.45), NIST (1.68, 4.01, 6.86, 9.18, 12.45)
ORP
- Range: ±2000mV
- Rel.mV Range: ±2000mV
- Resolution: 0.1mV (±199.9mV) / 1mV (beyond)
- Accuracy: ±0.2mV (±199.9mV) / ±2mV (beyond)
Temperature
- Range (meter): 0.0 to 100.0 °C / 32.0 to 212.0 °F
- Resolution: 0.1 °C / 0.1 °F
- Accuracy: ±0.3 °C / ±0.5 °F (0 to 70 °C)
- Compensation: ATC /mTC (0 to 100 °C) (pH only)
- Calibration: Offset in 0.1 °increments; offset range: ±5.0 °C / ±9.0 °F
- Power Requirements: 9 V DC adapter, 1.3 A (100/240 VAC, SmPS)
Spezifikation
Eutech pH 700 Meter With pH Electrode (ECFC7252101B), ATC Probe (PH5TEMB01P), Integral Electrode Holder and 100/240 VAC Adapter | |
±0.2 (±199.9), ±2 (Beyond) | |
0.01 | |
0°C to 100°C |
±0.01 | |
0.3°C/0.5°F | |
Up to 5 |